accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59-46-1

IUPAC Name: 2-(diethylamino)ethyl 4-aminobenzoate
Synonyms: procaine jenacaine lophakomp-procain n lophakomp-procain N stoff H3 base procaine procaina allocaine jenacain procain-loges
Molecular Weight (g/mol): 236.32
Molecular Formula: C13H20N2O2
InChi Key: MFDFERRIHVXMIY-UHFFFAOYSA-N
SMILES: CCN(CC)CCOC(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results