accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59-49-4

IUPAC Name: 2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: O-phenylene carbamate
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: ASSKVPFEZFQQNQ-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2O1
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)