accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNH2OHO

CAS RN 59-51-8

IUPAC Name: 2-amino-4-(methylsulfanyl)butanoic acid
Synonyms: methionin|metionina|Hmet
Molecular Weight (g/mol): 149.21
Molecular Formula: C5H11NO2S
InChi Key: FFEARJCKVFRZRR-UHFFFAOYNA-N
SMILES: CSCCC(N)C(O)=O
Percent Purity 
  • (2)
  • (4)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (4)
  • (2)
  • (7)
  • (4)
  • (2)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)