accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59-87-0

IUPAC Name: [(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea
Synonyms: nitrofurazone monofuracin cocafurin furesol furatsilin nitrofurazan sanfuran babrocid actin-N Furacin-HC
Molecular Weight (g/mol): 198.14
Molecular Formula: C6H6N4O4
InChi Key: IAIWVQXQOWNYOU-FPYGCLRLSA-N
SMILES: NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)