accessibility menu, dialog, popup

UserName

Viewing profile as user:
KCH3OO(E)(E)

CAS RN 590-00-1

IUPAC Name: potassium (2E,4E)-hexa-2,4-dienoate
Synonyms: potassium sorbate
Molecular Weight (g/mol): 150.22
Molecular Formula: C6H7KO2
InChi Key: CHHHXKFHOYLYRE-STWYSWDKSA-M
SMILES: [K+].C\C=C\C=C\C([O-])=O
Grade 
  • (5)
  • (5)
Percent Purity 
  • (10)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (10)
  • (10)
  • (10)
  • (5)
  • (5)
  • (10)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (4)