accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C(Z)

CAS RN 590-18-1

IUPAC Name: (2Z)-but-2-ene
Synonyms: cis-2-butene high-boiling butene-2
Molecular Weight (g/mol): 56.11
Molecular Formula: C4H8
InChi Key: IAQRGUVFOMOMEM-ARJAWSKDSA-N
SMILES: C\C=C/C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results