accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 590-28-3

IUPAC Name: potassium cyanate
Synonyms: potassium cyanate
Molecular Weight (g/mol): 81.12
Molecular Formula: CKNO
InChi Key: GKKCIDNWFBPDBW-UHFFFAOYSA-M
SMILES: [K+].[O-]C#N
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)