accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NCS

CAS RN 590-42-1

IUPAC Name: 2-isothiocyanato-2-methylpropane
Synonyms: 2-isothiocyanato-2-methylpropane
Molecular Weight (g/mol): 115.19
Molecular Formula: C5H9NS
InChi Key: ZFWFRTVIIMTOLY-UHFFFAOYSA-N
SMILES: CC(C)(C)N=C=S
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)