accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3H3C

CAS RN 590-73-8

IUPAC Name: 2,2-dimethylhexane
Synonyms: 2,2-dimethylhexane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: FLTJDUOFAQWHDF-UHFFFAOYSA-N
SMILES: CCCCC(C)(C)C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results