accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3O

CAS RN 590-86-3

IUPAC Name: 3-methylbutanal
Synonyms: isovaleraldehyde 3-methylbutanal (substance) aldehyde isovalerianique isovalerylaldehyde
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: YGHRJJRRZDOVPD-UHFFFAOYSA-N
SMILES: CC(C)CC=O
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (7)
Boiling Point 
  • (3)
  • (3)
  • (4)
  • (3)
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (7)
  • (1)
  • (1)
  • (3)
  • (3)
  • (7)
  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
  • (1)