accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHO

CAS RN 590-90-9

IUPAC Name: 4-hydroxybutan-2-one
Synonyms: 4-hydroxy-2-butanone
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: LVSQXDHWDCMMRJ-UHFFFAOYSA-N
SMILES: CC(=O)CCO
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)