accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOClH2N

CAS RN 5900-58-3

IUPAC Name: methyl 2-amino-4-chlorobenzoate
Synonyms: methyl 2-amino-4-chlorobenzoate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: YPSSCICDVDOEAI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Cl)C=C1N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)