accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SnH3CCH3N

CAS RN 59020-06-3

IUPAC Name: 4-(trimethylstannyl)pyridine
Synonyms: 4-(trimethylstannyl)pyridine
Molecular Weight (g/mol): 241.91
Molecular Formula: C8H13NSn
InChi Key: YCBWXJZXFUKDPU-UHFFFAOYSA-N
SMILES: C[Sn](C)(C)C1=CC=NC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results