accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSO

CAS RN 59020-90-5

IUPAC Name: {[(furan-2-yl)methyl]sulfanyl}carbaldehyde
Synonyms: (furan-2-ylmethyl)sulfanylcarbaldehyde
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: GFAOAYJTEVHTLA-UHFFFAOYSA-N
SMILES: O=CSCC1=CC=CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results