accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOO

CAS RN 59025-03-5

IUPAC Name: 2,5-dioxooxolan-3-yl acetate
Synonyms: 2,5-dioxooxolan-3-yl acetate
Molecular Weight (g/mol): 158.11
Molecular Formula: C6H6O5
InChi Key: SSWJHSASZZAIAU-UHFFFAOYNA-N
SMILES: CC(=O)OC1CC(=O)OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results