accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59026-32-3

IUPAC Name: 5-ethyl-5-(4-methylphenyl)-1,3-diazinane-4,6-dione
Synonyms: 5-ethyl-5-(4-methylphenyl)-1,3-diazinane-4,6-dione
Molecular Weight (g/mol): 232.28
Molecular Formula: C13H16N2O2
InChi Key: QEZWGNXBAXQVJT-UHFFFAOYSA-N
SMILES: CCC1(C(=O)NCNC1=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results