accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNBr

CAS RN 590417-95-1

IUPAC Name: 6-bromo-2-methyl-2H-indazole
Synonyms: 6-bromo-2-methylindazole
Molecular Weight (g/mol): 211.06
Molecular Formula: C8H7BrN2
InChi Key: BVYFYDANLZQCPV-UHFFFAOYSA-N
SMILES: CN1C=C2C=CC(Br)=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results