accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2N

CAS RN 5906-35-4

IUPAC Name: azepan-1-amine
Synonyms: azepan-1-amine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: UGBKOURNNQREPE-UHFFFAOYSA-N
SMILES: NN1CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results