accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59089-89-3

IUPAC Name: 2H,5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiine-2-thione
Synonyms: 5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiine-2-thione
Molecular Weight (g/mol): 224.39
Molecular Formula: C5H4S5
InChi Key: LFPIFRGVCYRUEO-UHFFFAOYSA-N
SMILES: S=C1SC2=C(S1)SCCS2
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results