accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 591-19-5

IUPAC Name: 3-bromoaniline
Synonyms: M-bromoaniline
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: DHYHYLGCQVVLOQ-UHFFFAOYSA-N
SMILES: NC1=CC=CC(Br)=C1
Boiling Point 
  • (8)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (8)
  • (2)
  • (2)
  • (8)
  • (2)
  • (8)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)