accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 591-60-6

IUPAC Name: butyl 3-oxobutanoate
Synonyms: butyl 3-oxobutanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: REIYHFWZISXFKU-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results