accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 591-93-5

IUPAC Name: penta-1,4-diene
Synonyms: 1,4-pentadiene
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: QYZLKGVUSQXAMU-UHFFFAOYSA-N
SMILES: C=CCC=C
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)