accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 591-95-7

IUPAC Name: penta-1,2-diene
Synonyms: 1,2-pentadiene
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: LVMTVPFRTKXRPH-UHFFFAOYNA-N
SMILES: CCC=C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results