CAS RN 5910-25-8
IUPAC Name:
3-phenylpentane-2,4-dione
Synonyms:
3-phenylpentane-2,4-dione
Molecular Weight (g/mol):
176.22
Molecular Formula:
C11H12O2
InChi Key:
YIWTXSVNRCWBAC-UHFFFAOYSA-N
SMILES:
CC(=O)C(C(C)=O)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Phenyl-2,4-pentanedione 98.0+%, TCI America™
CAS: 5910-25-8 Molecular Formula: C11H12O2 Molecular Weight (g/mol): 176.215 MDL Number: MFCD00048223 InChI Key: YIWTXSVNRCWBAC-UHFFFAOYSA-N Synonym: 3-Phenylacetylacetone PubChem CID: 233402 IUPAC Name: 3-phenylpentane-2,4-dione SMILES: CC(=O)C(C1=CC=CC=C1)C(=O)C