accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3C

CAS RN 5910-89-4

IUPAC Name: 2,3-dimethylpyrazine
Synonyms: pyrazine, 2,3-dimethyl-
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: OXQOBQJCDNLAPO-UHFFFAOYSA-N
SMILES: CC1=NC=CN=C1C
Boiling Point 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)