accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5911-04-6

H3CCH3CH3

CAS RN 5911-04-6

IUPAC Name: 3-methylnonane
Synonyms: nonane, 3-methyl-
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: PLZDDPSCZHRBOY-UHFFFAOYNA-N
SMILES: CCCCCCC(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results