accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59128-04-0

IUPAC Name: ethyl 2-{6-bromoimidazo[1,2-a]pyridin-2-yl}acetate
Synonyms: ethyl 2-{6-bromoimidazo[1,2-a]pyridin-2-yl}acetate
Molecular Weight (g/mol): 283.13
Molecular Formula: C11H11BrN2O2
InChi Key: VBLCCEAJXYZCRL-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CN2C=C(Br)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results