accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59142-68-6

IUPAC Name: 2-bromo-4-fluorobenzaldehyde
Synonyms: 2-bromo-4-fluorobenzaldehyde
Molecular Weight (g/mol): 203.01
Molecular Formula: C7H4BrFO
InChi Key: OPZDXMCOWFPQPE-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results