accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNClNHNCH3H3CCH3N

CAS RN 5915-41-3

IUPAC Name: N2-tert-butyl-6-chloro-N4-ethyl-1,3,5-triazine-2,4-diamine
Synonyms: terbuthylazine chlorcaragard epa pesticide chemical code 080814 gardeprim A 1862 terbutylethylazine primatol-M80 terbythylazine terbutylazine caswell no. 125B gardoprim
Molecular Weight (g/mol): 229.71
Molecular Formula: C9H16ClN5
InChi Key: FZXISNSWEXTPMF-UHFFFAOYSA-N
SMILES: CCNC1=NC(Cl)=NC(NC(C)(C)C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results