accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSiClClCl

CAS RN 592-09-6

IUPAC Name: trichloro(3,3,3-trifluoropropyl)silane
Synonyms: trichloro(3,3,3-trifluoropropyl)silane
Molecular Weight (g/mol): 231.5
Molecular Formula: C3H4Cl3F3Si
InChi Key: WEUBQNJHVBMUMD-UHFFFAOYSA-N
SMILES: FC(F)(F)CC[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results