accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CN

CAS RN 592-51-8

IUPAC Name: pent-4-enenitrile
Synonyms: 4-pentenenitrile
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: CFEYBLWMNFZOPB-UHFFFAOYSA-N
SMILES: C=CCCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results