accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 592-57-4

IUPAC Name: cyclohexa-1,3-diene
Synonyms: 1,3-cyclohexadiene
Molecular Weight (g/mol): 80.13
Molecular Formula: C6H8
InChi Key: MGNZXYYWBUKAII-UHFFFAOYSA-N
SMILES: C1CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)