accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 592-65-4

IUPAC Name: 2-methyl-1-[(2-methylpropyl)sulfanyl]propane
Synonyms: 2-methyl-1-[(2-methylpropyl)sulfanyl]propane
Molecular Weight (g/mol): 146.29
Molecular Formula: C8H18S
InChi Key: CMWSRWTXVQLHNX-UHFFFAOYSA-N
SMILES: CC(C)CSCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results