accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 592-78-9

IUPAC Name: (3E)-hept-3-ene
Synonyms: 3-heptene (cis) 3-heptene (trans)
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: WZHKDGJSXCTSCK-FNORWQNLSA-N
SMILES: CCC\C=C\CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results