accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 592-84-7

IUPAC Name: butyl formate
Synonyms: butyl formate N-butylformate
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: NMJJFJNHVMGPGM-UHFFFAOYSA-N
SMILES: CCCCOC=O
Boiling Point 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)