accessibility menu, dialog, popup

UserName

Viewing profile as user:
PbSNSN

CAS RN 592-87-0

IUPAC Name: λ²-lead(2+) bis(cyanosulfanide)
Synonyms: lead(II) thiocyanate
Molecular Weight (g/mol): 323.4
Molecular Formula: C2N2PbS2
InChi Key: VRNINGUKUJWZTH-UHFFFAOYSA-L
SMILES: [Pb++].[S-]C#N.[S-]C#N
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)