accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 592-88-1

IUPAC Name: 3-(prop-2-en-1-ylsulfanyl)prop-1-ene
Synonyms: oil garlic allium sativum oil oil of garlic (substance) thioallyl ether garlic oil oil of garlic garlic oil preparation
Molecular Weight (g/mol): 114.21
Molecular Formula: C6H10S
InChi Key: UBJVUCKUDDKUJF-UHFFFAOYSA-N
SMILES: C=CCSCC=C
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)