accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59280-70-5

IUPAC Name: N-(4-chloro-2-fluorophenyl)acetamide
Synonyms: 2-fluoro-4-chloroacetanilide
Molecular Weight (g/mol): 187.6
Molecular Formula: C8H7ClFNO
InChi Key: GVRKNSAEOVXHOS-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Cl)C=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results