accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 593-82-8

HClCH3NCH3H2N

CAS RN 593-82-8

IUPAC Name: hydrogen 1,1-dimethylhydrazine chloride
Synonyms: hydrogen 1,1-dimethylhydrazine chloride
Molecular Weight (g/mol): 96.56
Molecular Formula: C2H9ClN2
InChi Key: VFQADAFGYKTPSH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results