accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5931-17-9

IUPAC Name: 4-({[(4-hydroxybutyl)dimethylsilyl]oxy}dimethylsilyl)butan-1-ol
Synonyms: 4-({[(4-hydroxybutyl)dimethylsilyl]oxy}dimethylsilyl)butan-1-ol
Molecular Weight (g/mol): 278.54
Molecular Formula: C12H30O3Si2
InChi Key: OWJKJLOCIDNNGJ-UHFFFAOYSA-N
SMILES: C[Si](C)(CCCCO)O[Si](C)(C)CCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results