CAS RN 5932-27-4
IUPAC Name:
1-{4-[(2-chlorophenyl)methyl]piperazin-1-yl}-3-methylbutan-1-one
Synonyms:
1-{4-[(2-chlorophenyl)methyl]piperazin-1-yl}-3-methylbutan-1-one
Molecular Weight (g/mol):
294.82
Molecular Formula:
C16H23ClN2O
InChi Key:
WENGXLQDBFYGRD-UHFFFAOYSA-N
SMILES:
CC(C)CC(=O)N1CCN(CC2=CC=CC=C2Cl)CC1
1
–
1
of
1
results
1
Ethyl Pyrazole-3-carboxylate 98.0+%, TCI America™
CAS: 5932-27-4 Molecular Formula: C6H8N2O2 Molecular Weight (g/mol): 140.142 MDL Number: MFCD00159643 InChI Key: MSPOSRHJXMILNK-UHFFFAOYSA-N Synonym: Pyrazole-3-carboxylic Acid Ethyl Ester PubChem CID: 7147518 IUPAC Name: ethyl 1H-pyrazole-5-carboxylate SMILES: CCOC(=O)C1=CC=NN1