accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOClNOO

CAS RN 59320-12-6

IUPAC Name: N-butyl-4-chloro-3-nitrobenzamide
Synonyms: N-butyl-4-chloro-3-nitrobenzamide
Molecular Weight (g/mol): 256.69
Molecular Formula: C11H13ClN2O3
InChi Key: YVBGUSUYCUSCBC-UHFFFAOYSA-N
SMILES: CCCCNC(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results