accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSCl

CAS RN 59337-89-2

IUPAC Name: 3-chlorothiophene-2-carboxylate
Synonyms: 3-chlorothiophene-2-carboxylate
Molecular Weight (g/mol): 161.58
Molecular Formula: C5H2ClO2S
InChi Key: BXEAAHIHFFIMIE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(Cl)C=CS1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)