accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOOOOHOOOOHOKKKFe

CAS RN 5936-11-8

IUPAC Name: tripotassium iron trihydrogen oxalate
Synonyms: tripotassium iron trihydrogen oxalate
Molecular Weight (g/mol): 440.22
Molecular Formula: C6H3FeK3O12
InChi Key: YXTWCYYDGRBUDE-UHFFFAOYSA-K
SMILES: [K+].[K+].[K+].[Fe].OC(=O)C([O-])=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)