accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59376-58-8

IUPAC Name: (2Z,4Z)-undeca-2,4-dien-1-ol
Synonyms: (2Z,4Z)-undeca-2,4-dien-1-ol
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: FVKXLSPKNRZPJK-QRLRYFCNSA-N
SMILES: CCCCCC\C=C/C=C\CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results