accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59382-39-7

IUPAC Name: 1-fluoro-4-iodo-2-(trifluoromethyl)benzene
Synonyms: 1-fluoro-4-iodo-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 290
Molecular Formula: C7H3F4I
InChi Key: DKLKYTATXLQGMX-UHFFFAOYSA-N
SMILES: FC1=C(C=C(I)C=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results