accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59388-58-8

CH3H3CH3CHNCH3

CAS RN 59388-58-8

IUPAC Name: 2,2,4,7-tetramethyl-1,2,3,4-tetrahydroquinoline
Synonyms: 2,2,4,7-tetramethyl-3,4-dihydro-1H-quinoline
Molecular Weight (g/mol): 189.3
Molecular Formula: C13H19N
InChi Key: NRWNXIXJZMSDAU-UHFFFAOYNA-N
SMILES: CC1CC(C)(C)NC2=CC(C)=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results