accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 594-19-4

IUPAC Name: tert-butyllithium
Synonyms: tert-butyllithium
Molecular Weight (g/mol): 64.06
Molecular Formula: C4H9Li
InChi Key: BKDLGMUIXWPYGD-UHFFFAOYSA-N
SMILES: [Li]C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)