accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 594-20-7

IUPAC Name: 2,2-dichloropropane
Synonyms: propane, 2,2-dichloro-
Molecular Weight (g/mol): 112.98
Molecular Formula: C3H6Cl2
InChi Key: ZEOVXNVKXIPWMS-UHFFFAOYSA-N
SMILES: CC(C)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)