accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3OO

CAS RN 594-43-4

IUPAC Name: methanesulfonylethane
Synonyms: methanesulfonylethane
Molecular Weight (g/mol): 108.16
Molecular Formula: C3H8O2S
InChi Key: YBJCDTIWNDBNTM-UHFFFAOYSA-N
SMILES: CCS(C)(=O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)